Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxw15
Entrez Gene ID
382105
Gene Name
F-box and WD-40 domain protein 15
Gene Aliases
Fbxo12J
Chromosome Location
Chr.9: 109552602 - 109569438 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199036.2 NP_950201.2 9 - 10 1324 75
GenBank mRNA AK087669.1 - 9 - 10 1325 75
BC099378.1 - 6 - 7 763 75
CO795020.1 - 3 - 4 345 75
DQ067445.1 - 9 - 10 1292 75

View Full Product Details