The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111304.1 | NP_001104774.1 | 13 - 14 | 3058 | 60 |
NM_027758.4 | NP_082034.1 | 10 - 11 | 2359 | 60 | |
GenBank mRNA | AK049049.1 | - | 10 - 11 | 2360 | 60 |
AK134614.1 | - | 13 - 14 | 2335 | 60 | |
AK173050.1 | - | 13 - 14 | 2231 | 60 | |
BC057027.1 | - | 9 - 10 | 1646 | 60 |