Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d9
Entrez Gene ID
71310
Gene Name
TBC1 domain family, member 9
Gene Aliases
4933431N12Rik, AI847101, AW490653, C76116
Chromosome Location
Chr.8: 83165352 - 83272940 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111304.1 NP_001104774.1 1 - 2 855 109
NM_027758.4 NP_082034.1 1 - 2 855 109
GenBank mRNA AK049049.1 - 1 - 2 856 109
AK134614.1 - 1 - 2 132 109

View Full Product Details