The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039154.2 | NP_001034243.1 | 1 - 2 | 390 | 77 |
NM_001285913.1 | NP_001272842.1 | 1 - 2 | 390 | 77 | |
NM_001285914.1 | NP_001272843.1 | 1 - 2 | 390 | 77 | |
NM_007667.3 | NP_031693.2 | 1 - 2 | 390 | 77 | |
GenBank mRNA | AK041720.1 | - | 1 - 2 | 358 | 77 |
AK044046.1 | - | 1 - 2 | 347 | 77 | |
AK083092.1 | - | 1 - 2 | 357 | 77 | |
AK086711.1 | - | 1 - 2 | 359 | 77 | |
AK139334.1 | - | 1 - 2 | 346 | 77 | |
BC057581.1 | - | 1 - 2 | 295 | 77 | |
BC060200.1 | - | 1 - 2 | 238 | 77 | |
BU058167.1 | - | 1 - 2 | 390 | 77 |