Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jph3
Entrez Gene ID
57340
Gene Name
junctophilin 3
Gene Aliases
JP-3, Jp3
Chromosome Location
Chr.8: 121730559 - 121791083 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020605.3 NP_065630.1 1 - 2 446 83
GenBank mRNA AB024449.1 - 1 - 2 442 83
AK045706.1 - 1 - 2 433 83
AK046136.1 - - 439 83
AK082880.1 - 1 - 2 435 83
BC103682.1 - 1 - 2 401 83
BC104736.1 - 1 - 2 401 83
BC105307.1 - 1 - 2 401 83
BY718602.1 - 1 - 2 457 83

View Full Product Details