Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctu2
Entrez Gene ID
66965
Gene Name
cytosolic thiouridylase subunit 2 homolog (S. pombe)
Gene Aliases
2310061F22Rik, C16orf84, Ncs2
Chromosome Location
Chr.8: 122476137 - 122483092 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153775.2 NP_722470.2 5 - 6 428 67
GenBank mRNA AK012440.1 - 5 - 6 66
AK090391.1 - - 363 67
AK154995.1 - 5 - 6 363 67
AK172353.1 - 3 - 4 871 67
BC036332.1 - 5 - 6 381 67

View Full Product Details