Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd13b
Entrez Gene ID
268445
Gene Name
ankyrin repeat domain 13b
Gene Aliases
AW124583, B930093C12Rik
Chromosome Location
Chr.11: 77470485 - 77489678 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172945.2 NP_766533.2 5 - 6 692 68
GenBank mRNA BC151158.1 - 5 - 6 670 68
BC172028.1 - 5 - 6 670 68
CA315262.1 - 4 - 5 378 68
CA329030.1 - 5 - 6 680 68

View Full Product Details