Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd13b
Entrez Gene ID
268445
Gene Name
ankyrin repeat domain 13b
Gene Aliases
AW124583, B930093C12Rik
Chromosome Location
Chr.11: 77470485 - 77489678 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172945.2 NP_766533.2 10 - 11 1231 118
GenBank mRNA BC056928.1 - 5 - 6 460 118
BC071189.1 - 4 - 5 317 118
BC098207.1 - 3 - 4 270 118
BC151158.1 - 10 - 11 1209 118
BC172028.1 - 10 - 11 1210 118
CD578820.1 - 4 - 5 329 118

View Full Product Details