Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gse1
Entrez Gene ID
382034
Gene Name
genetic suppressor element 1
Gene Aliases
2210013I18Rik, mKIAA0182
Chromosome Location
Chr.8: 120228826 - 120581383 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145896.1 NP_001139368.1 4 - 5 744 107
NM_001145897.1 NP_001139369.1 4 - 5 688 107
NM_198671.2 NP_941073.2 4 - 5 876 107
GenBank mRNA AK033263.1 - 4 - 5 691 107
AK042915.1 - 4 - 5 692 107
AK154829.1 - 4 - 5 723 107
BC043094.1 - 4 - 5 876 107

View Full Product Details