The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130187.1 | NP_001123659.1 | 1 - 2 | 477 | 75 |
GenBank mRNA | AK159259.1 | - | 1 - 2 | 314 | 75 |
AK166123.1 | - | 1 - 2 | 270 | 75 |