Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nup85
Entrez Gene ID
445007
Gene Name
nucleoporin 85
Gene Aliases
Pcnt1, frount
Chromosome Location
Chr.11: 115564432 - 115583924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002929.4 NP_001002929.3 6 - 7 567 82
GenBank mRNA AF498263.1 - 6 - 7 481 82
AK017632.1 - 6 - 7 532 82
BC079856.1 - 6 - 7 542 82

View Full Product Details