The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054046.5 | NP_473387.1 | 1 - 2 | 93 | 66 |
GenBank mRNA | AK034028.1 | - | - | 59 | 66 |
AK046271.1 | - | 1 - 2 | 54 | 66 | |
AK086174.1 | - | 1 - 2 | 54 | 66 | |
AK087778.1 | - | 1 - 2 | 64 | 66 | |
AK133979.1 | - | 1 - 2 | 67 | 66 | |
AK164147.1 | - | 1 - 2 | 65 | 66 |