Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc38a7
Entrez Gene ID
234595
Gene Name
solute carrier family 38, member 7
Gene Aliases
D430050E18
Chromosome Location
Chr.8: 95835920 - 95853577 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172758.4 NP_766346.1 5 - 6 860 87
GenBank mRNA AK052548.1 - 5 - 6 861 87
AK171137.1 - 5 - 6 868 87
BC031853.1 - 3 - 4 735 87

View Full Product Details