Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wfdc16
Entrez Gene ID
277345
Gene Name
WAP four-disulfide core domain 16
Gene Aliases
BC043944, WAP16
Chromosome Location
Chr.2: 164634701 - 164638808 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012723.2 NP_001012741.2 2 - 3 214 127
GenBank mRNA AY542488.1 - 2 - 3 214 127
AY542489.1 - 2 - 3 214 127
BC043944.1 - 2 - 3 208 127
BC145970.1 - 1 - 2 161 127
BC145972.1 - 1 - 2 161 127

View Full Product Details