The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024191.2 | NP_077153.1 | 1 - 2 | 305 | 72 |
GenBank mRNA | AK018229.1 | - | 1 - 2 | 115 | 72 |
AK149620.1 | - | 1 - 2 | 143 | 72 | |
AK155242.1 | - | 1 - 2 | 256 | 72 | |
AK161325.1 | - | 1 - 2 | 124 | 72 | |
BC002131.1 | - | 1 - 2 | 150 | 72 | |
BC024708.1 | - | 1 - 2 | 254 | 72 | |
CT010168.1 | - | 1 - 2 | 41 | 72 |