The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001333.2 | NP_001001333.2 | 12 - 13 | 1715 | 75 |
NM_001146073.1 | NP_001139545.1 | 11 - 12 | 1520 | 75 | |
GenBank mRNA | AK154237.1 | - | 12 - 13 | 1689 | 75 |
BC056487.1 | - | 8 - 9 | 3485 | 75 | |
BC069960.1 | - | 11 - 12 | 1472 | 75 | |
FM204885.1 | - | 11 - 12 | 1252 | 75 | |
FM204886.1 | - | 11 - 12 | 1471 | 75 |