The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009204.2 | NP_033230.2 | 10 - 11 | 1537 | 65 |
GenBank mRNA | AB008453.1 | - | 10 - 11 | 1335 | 65 |
AK137607.1 | - | 10 - 11 | 1522 | 65 | |
AK160882.1 | - | 8 - 9 | 2162 | 65 | |
AK164310.1 | - | 10 - 11 | 1512 | 65 | |
BC014282.1 | - | 10 - 11 | 1507 | 65 |