Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Asgr2
Entrez Gene ID
11890
Gene Name
asialoglycoprotein receptor 2
Gene Aliases
ASGPR2, Asgr, Asgr-2
Chromosome Location
Chr.11: 70092307 - 70106187 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007493.3 NP_031519.1 6 - 7 656 57
GenBank mRNA BC011197.1 - 6 - 7 656 57
X53042.1 - 6 - 7 532 57

View Full Product Details