Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pelp1
Entrez Gene ID
75273
Gene Name
proline, glutamic acid and leucine rich protein 1
Gene Aliases
4930563C04Rik, MNAR
Chromosome Location
Chr.11: 70392881 - 70410031 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029231.4 NP_083507.3 9 - 10 1091 75
GenBank mRNA AK005027.1 - 9 - 10 1089 75
AK128966.1 - 9 - 10 1091 75
BC057987.1 - 3 - 4 319 75
BC090620.1 - 9 - 10 1085 75

View Full Product Details