The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033794.3 | NP_001028966.1 | 3 - 4 | 351 | 82 |
GenBank mRNA | AK136230.1 | - | 3 - 4 | 355 | 82 |
AK162167.1 | - | 3 - 4 | 336 | 82 | |
BC139081.1 | - | 3 - 4 | 351 | 82 |