Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wscd1
Entrez Gene ID
216881
Gene Name
WSC domain containing 1
Gene Aliases
mKIAA0523
Chromosome Location
Chr.11: 71749923 - 71789647 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177618.4 NP_808286.2 5 - 6 1232 109
GenBank mRNA AK220346.1 - 4 - 5 938 109
BC030477.1 - 5 - 6 1369 109
BC048831.1 - 4 - 5 1051 109
BC063070.1 - 5 - 6 1232 109

View Full Product Details