Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Serpinf2
Entrez Gene ID
18816
Gene Name
serine (or cysteine) peptidase inhibitor, clade F, member 2
Gene Aliases
AI747498, Pli, Serpimf2
Chromosome Location
Chr.11: 75431736 - 75439501 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008878.2 NP_032904.1 4 - 5 208 62
GenBank mRNA AK077563.1 - 4 - 5 211 62
AK090208.1 - 4 - 5 192 62
BC026756.1 - 4 - 5 182 62
GQ896353.1 - 3 - 4 159 62
GQ896354.1 - 3 - 4 159 62
Z36774.1 - 4 - 5 195 62

View Full Product Details