Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankfy1
Entrez Gene ID
11736
Gene Name
ankyrin repeat and FYVE domain containing 1
Gene Aliases
Ankhzn, ZFYVE14, mKIAA1255
Chromosome Location
Chr.11: 72690002 - 72772146 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009671.5 NP_033801.4 9 - 10 1187 75
GenBank mRNA AB098329.1 - 9 - 10 1185 75
BC139231.1 - 9 - 10 1198 75
BC145583.1 - 9 - 10 1198 75
CV557421.1 - 2 - 3 214 75

View Full Product Details