Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcakd
Entrez Gene ID
68087
Gene Name
dephospho-CoA kinase domain containing
Gene Aliases
3010024O21Rik, 6720485C15Rik, AI849483
Chromosome Location
Chr.11: 102994047 - 103028239 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026551.3 NP_080827.2 1 - 2 102 103
GenBank mRNA AK051022.1 - 1 - 2 103 103
AK162069.1 - 1 - 2 62 103
BC005607.1 - 1 - 2 68 103
BC045193.1 - 1 - 2 56 103

View Full Product Details