Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyb5d2
Entrez Gene ID
192986
Gene Name
cytochrome b5 domain containing 2
Gene Aliases
9330151E16Rik, Gm2
Chromosome Location
Chr.11: 72777232 - 72795839 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024926.3 NP_001020097.2 3 - 4 658 80
GenBank mRNA AK034043.1 - 3 - 4 356 80
BC086682.1 - 3 - 4 624 80
BC117721.1 - 2 - 3 240 80

View Full Product Details