Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ube2o
Entrez Gene ID
217342
Gene Name
ubiquitin-conjugating enzyme E2O
Gene Aliases
9630022H21, B230113M03Rik, E2-230K, mKIAA1734
Chromosome Location
Chr.11: 116537740 - 116581476 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173755.3 NP_776116.2 10 - 11 1806 95
GenBank mRNA AK129431.1 - 10 - 11 1806 95
BC039233.1 - 2 - 3 272 95
BC059193.1 - 10 - 11 1431 95
U20780.1 - 10 - 11 1467 95

View Full Product Details