The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178645.4 | NP_848760.1 | 5 - 6 | 783 | 75 |
GenBank mRNA | AK029274.1 | - | 4 - 5 | 1676 | 75 |
AK035228.1 | - | 5 - 6 | 657 | 75 | |
AK040799.1 | - | 5 - 6 | 673 | 75 | |
AK049461.1 | - | 5 - 6 | 695 | 75 | |
AK051441.1 | - | 5 - 6 | 682 | 75 | |
AK053872.1 | - | 5 - 6 | 663 | 75 | |
AK150438.1 | - | 5 - 6 | 682 | 75 | |
AK150896.1 | - | 5 - 6 | 663 | 75 | |
AK167325.1 | - | 5 - 6 | 663 | 75 | |
BC024090.1 | - | 5 - 6 | 668 | 75 | |
BC027037.1 | - | 5 - 6 | 639 | 75 | |
BC027362.1 | - | 5 - 6 | 672 | 75 | |
BC027403.1 | - | 5 - 6 | 693 | 75 |