Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atxn7l3
Entrez Gene ID
217218
Gene Name
ataxin 7-like 3
Gene Aliases
E030022H21Rik
Chromosome Location
Chr.11: 102289300 - 102296643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098837.1 NP_001092307.1 6 - 7 769 101
GenBank mRNA AK090368.1 - 6 - 7 771 101

View Full Product Details