Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkar1a
Entrez Gene ID
19084
Gene Name
protein kinase, cAMP dependent regulatory, type I, alpha
Gene Aliases
1300018C22Rik, RIalpha, Tse-1, Tse1
Chromosome Location
Chr.11: 109649395 - 109669659 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021880.2 NP_068680.1 4 - 5 568 85
GenBank mRNA AK005039.1 - 4 - 5 570 85
AK019147.1 - 2 - 3 203 85
AK027916.1 - 4 - 5 535 85
AK049832.1 - 4 - 5 561 85
AK051068.1 - 4 - 5 569 85
AK053918.1 - 4 - 5 569 85
AK083383.1 - 4 - 5 549 85
AK145797.1 - 4 - 5 569 85
AK145860.1 - 4 - 5 536 85
AK146380.1 - 4 - 5 555 85
AK147188.1 - 4 - 5 560 85
AK150427.1 - 4 - 5 561 85
AK150927.1 - 2 - 3 201 85
AK151040.1 - 2 - 3 201 85
AK151124.1 - 4 - 5 527 85
AK151556.1 - 2 - 3 201 85
AK152044.1 - 2 - 3 201 85
AK153227.1 - 4 - 5 527 85
AK158562.1 - 4 - 5 906 85
BC003461.1 - 4 - 5 539 85
BC005697.1 - 4 - 5 510 85
HQ412666.1 - 5 - 6 751 85
HQ412667.1 - 4 - 5 567 85
HQ412668.1 - 4 - 5 627 85

View Full Product Details