The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252208.1 | NP_001239137.1 | 1 - 2 | 118 | 121 |
GenBank mRNA | BB850391.1 | - | 1 - 2 | 118 | 119 |