The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012046.2 | NP_036176.1 | 2 - 3 | 296 | 69 |
GenBank mRNA | AF169386.1 | - | 2 - 3 | 293 | 69 |
AF173848.1 | - | 2 - 3 | 289 | 69 |