The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007636.2 | NP_031662.2 | 14 - 15 | 1540 | 60 |
GenBank mRNA | AB041570.1 | - | 14 - 15 | 1371 | 60 |
AK012295.1 | - | 14 - 15 | 1500 | 60 | |
AK075581.1 | - | 14 - 15 | 1513 | 60 | |
AK088145.1 | - | 14 - 15 | 1502 | 60 | |
AK145406.1 | - | 14 - 15 | 1541 | 60 | |
AK166891.1 | - | 14 - 15 | 1513 | 60 | |
BC007470.1 | - | 14 - 15 | 1504 | 60 | |
BC026918.1 | - | 14 - 15 | 1506 | 60 | |
Z31553.1 | - | 14 - 15 | 1491 | 60 |