Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Zhx3
- Entrez Gene ID
-
320799
- Gene Name
- zinc fingers and homeoboxes 3
- Gene Aliases
-
1810059C13Rik, 4932418O04Rik, 9530010N21Rik, Tix1, mKIAA0395
- Chromosome Location
-
Chr.2: 160770447 - 160872990 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK030057.1 |
- |
1 - 2 |
161 |
70 |
View Full Product Details