The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253783.1 | NP_001240712.1 | 3 - 4 | 199 | 103 |
GenBank mRNA | BC003306.1 | - | 3 - 4 | 150 | 103 |