The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008524.2 | NP_032550.2 | 2 - 3 | 1274 | 135 |
GenBank mRNA | AK014312.1 | - | 2 - 3 | 970 | 135 |
AK075737.1 | - | 2 - 3 | 973 | 135 | |
AK085240.2 | - | 2 - 3 | 966 | 135 | |
AK164774.1 | - | 2 - 3 | 966 | 135 | |
BC005550.1 | - | 2 - 3 | 952 | 135 |