The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113486.1 | NP_001106958.1 | 11 - 12 | 1720 | 65 |
NM_001113487.1 | NP_001106959.1 | 10 - 11 | 1900 | 65 | |
NM_001113488.1 | NP_001106960.1 | 9 - 10 | 1069 | 65 | |
NM_017380.2 | NP_059076.1 | 9 - 10 | 1050 | 65 | |
GenBank mRNA | AJ250723.1 | - | 9 - 10 | 1056 | 65 |
AK031757.1 | - | 9 - 10 | 945 | 65 | |
AK122415.2 | - | 10 - 11 | 1694 | 65 | |
AK141312.1 | - | 11 - 12 | 1661 | 65 | |
AK164581.1 | - | 8 - 9 | 3664 | 65 | |
BC046524.1 | - | 11 - 12 | 1720 | 65 | |
BC094455.1 | - | 12 - 13 | 1424 | 65 |