Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttyh2
Entrez Gene ID
117160
Gene Name
tweety homolog 2 (Drosophila)
Gene Aliases
1110001A03Rik
Chromosome Location
Chr.11: 114675468 - 114720984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053273.2 NP_444503.2 11 - 12 1340 142
GenBank mRNA AF329682.1 - 11 - 12 1280 142
AK152172.1 - 11 - 12 1292 142
AK154835.1 - 11 - 12 1347 142
AK155166.1 - 11 - 12 1341 142
AK168408.1 - 11 - 12 1293 142
BC060740.1 - 11 - 12 1283 142

View Full Product Details