Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ube2v1
Entrez Gene ID
66589
Gene Name
ubiquitin-conjugating enzyme E2 variant 1
Gene Aliases
0610011J09Rik, AI256840, CROC-1, CROC1, D7Bwg1382e, UEV-1
Chromosome Location
Chr.2: 167607639 - 167632047 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023230.2 NP_075719.1 3 - 4 344 71
GenBank mRNA AK012021.1 - 3 - 4 323 71
AK158230.1 - 3 - 4 344 71
AK170413.1 - 3 - 4 315 71
BC003449.1 - 3 - 4 331 71
BC092253.1 - 3 - 4 310 71


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK079211.1 - Chr. - - [-] on Build 71

View Full Product Details