The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242558.1 | NP_001229487.1 | 1 - 2 | 98 | 113 |
NM_019825.3 | NP_062799.2 | 1 - 2 | 98 | 113 | |
GenBank mRNA | AK144338.1 | - | 3 - 4 | 337 | 113 |
BC031113.1 | - | 1 - 2 | 102 | 113 | |
BC083187.1 | - | 1 - 2 | 98 | 113 |