Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Susd5
Entrez Gene ID
382111
Gene Name
sushi domain containing 5
Gene Aliases
Gm1126
Chromosome Location
Chr.9: 114057354 - 114098733 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101510.1 NP_001094980.1 4 - 5 783 87
GenBank mRNA AK132299.1 - 4 - 5 88

View Full Product Details