The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027186.2 | NP_081462.1 | 5 - 6 | 793 | 71 |
GenBank mRNA | AK010289.1 | - | 5 - 6 | 522 | 71 |
BC047996.1 | - | 5 - 6 | 780 | 71 | |
DQ810267.1 | - | 5 - 6 | 486 | 71 | |
DQ810270.1 | - | 4 - 5 | 374 | 71 |