Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Itga2b
Entrez Gene ID
16399
Gene Name
integrin alpha 2b
Gene Aliases
AI172977, CD41, CD41B, GpIIb, alphaIIb
Chromosome Location
Chr.11: 102453297 - 102469883 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010575.2 NP_034705.2 10 - 11 1116 54
GenBank mRNA AF166384.1 - 10 - 11 994 54
AF170316.1 - 10 - 11 977 54
AK048700.1 - 6 - 7 471 54
AK140749.1 - 5 - 6 768 54
AK142289.1 - 10 - 11 978 54
AK154619.1 - 10 - 11 1117 54
AK156872.1 - 9 - 10 1298 54
AK158443.1 - 10 - 11 963 54
BC120493.1 - 10 - 11 966 54
BC137570.1 - 10 - 11 966 54

View Full Product Details