Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmd12
Entrez Gene ID
66997
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 12
Gene Aliases
1500002F15Rik, AI480719, P55
Chromosome Location
Chr.11: 107479528 - 107498036 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025894.2 NP_080170.1 10 - 11 1214 97
GenBank mRNA AK005114.1 - 10 - 11 1248 97
AK007619.1 - 10 - 11 1259 97
AK007680.1 - 10 - 11 1188 97
AK008646.1 - 10 - 11 1213 97
AK009969.1 - 10 - 11 1217 97
AK017843.1 - 10 - 11 1231 97
AK150910.1 - 10 - 11 1237 97
AK151242.1 - 10 - 11 1215 97
AK152041.1 - 10 - 11 1237 97
AK152240.1 - 10 - 11 1246 97
AK160283.1 - 10 - 11 1237 97
AK167236.1 - 10 - 11 1221 97
BC004694.1 - 10 - 11 1232 97

View Full Product Details