Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gan
Entrez Gene ID
209239
Gene Name
giant axonal neuropathy
Gene Aliases
A330045G18, gigaxonin
Chromosome Location
Chr.8: 117158135 - 117205186 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081151.1 NP_001074620.1 7 - 8 1243 71
GenBank mRNA AK039455.1 - 7 - 8 1242 71
BC112432.1 - 7 - 8 1242 71

View Full Product Details