Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dlgap4
Entrez Gene ID
228836
Gene Name
discs, large homolog-associated protein 4 (Drosophila)
Gene Aliases
AI225853, BC024558, DAP-4, DAP4, SAPAP-4, Sapap4, WBP16
Chromosome Location
Chr.2: 156613633 - 156764363 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042487.1 NP_001035952.1 6 - 7 1930 104
NM_001042488.2 NP_001035953.1 6 - 7 1229 104
NM_001277186.1 NP_001264115.1 12 - 13 3165 104
NM_001277187.1 NP_001264116.1 11 - 12 3123 104
NM_146128.6 NP_666240.4 12 - 13 3174 104
GenBank mRNA AK122408.1 - 10 - 11 5832 104
AK150554.1 - 6 - 7 1150 104
AK156212.1 - 4 - 5 3808 104
AY243849.2 - 10 - 11 2751 104
BC024558.1 - 6 - 7 1334 104
BC058948.1 - 6 - 7 1253 104
BC085475.1 - 6 - 7 1717 104
BC106094.1 - 6 - 7 1312 104
BC141110.1 - 10 - 11 2802 104
BC145394.1 - 10 - 11 2802 104
BC145395.1 - 9 - 10 2760 104

View Full Product Details