Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dlgap4
Entrez Gene ID
228836
Gene Name
discs, large homolog-associated protein 4 (Drosophila)
Gene Aliases
AI225853, BC024558, DAP-4, DAP4, SAPAP-4, Sapap4, WBP16
Chromosome Location
Chr.2: 156613633 - 156764363 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277186.1 NP_001264115.1 7 - 8 2069 121
NM_001277187.1 NP_001264116.1 6 - 7 2018 121
NM_146128.6 NP_666240.4 7 - 8 2069 121
GenBank mRNA AK122408.1 - 7 - 8 2069 121
AY243849.2 - 5 - 6 1649 121
BC141110.1 - 5 - 6 1706 121
BC145394.1 - 5 - 6 1706 121
BC145395.1 - 4 - 5 1655 121

View Full Product Details