The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172118.1 | NP_742116.1 | 1 - 2 | 47 | 59 |
GenBank mRNA | AK007972.1 | - | 1 - 2 | 58 | |
AK020258.1 | - | 1 - 2 | 48 | 59 | |
AK159880.1 | - | 1 - 2 | 92 | 59 | |
BC049974.1 | - | 1 - 2 | 82 | 59 | |
BC055439.1 | - | 1 - 2 | 71 | 59 |