Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Palm3
Entrez Gene ID
74337
Gene Name
paralemmin 3
Gene Aliases
4432412L15Rik, AL024300
Chromosome Location
Chr.8: 84021474 - 84030295 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028877.1 NP_083153.1 5 - 6 412 76
GenBank mRNA BC113196.1 - 5 - 6 415 76
BC113802.1 - 5 - 6 410 76
EF208034.1 - 5 - 6 412 76

View Full Product Details