The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252384.1 | NP_001239313.1 | 3 - 4 | 571 | 86 |
NM_001252394.1 | NP_001239323.1 | 2 - 3 | 408 | 86 | |
NM_001252395.1 | NP_001239324.1 | 2 - 3 | 408 | 86 | |
NM_026071.3 | NP_080347.1 | 2 - 3 | 408 | 86 | |
GenBank mRNA | AK005710.1 | - | 2 - 3 | 253 | 86 |
AK168145.1 | - | 2 - 3 | 207 | 86 | |
AK172622.1 | - | 1 - 2 | 210 | 86 | |
BC018167.1 | - | 2 - 3 | 214 | 86 | |
BC046767.1 | - | 2 - 3 | 258 | 86 | |
CX241396.1 | - | 3 - 4 | 377 | 86 |