The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025305.2 | NP_079581.1 | 2 - 3 | 283 | 68 |
GenBank mRNA | AK009251.1 | - | 2 - 3 | 248 | 68 |
AK012225.1 | - | 2 - 3 | 283 | 68 | |
AK131937.1 | - | 2 - 3 | 294 | 68 | |
AK162024.1 | - | 2 - 3 | 262 | 68 | |
BC049636.1 | - | 2 - 3 | 283 | 68 |